Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Armc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107306.1 | NP_001100776.1 | 4 | 1101 | ||
GenBank mRNA | BC166821.1 | - | 757 | 8368184 |