Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBL4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014235.4 | NP_055050.1 | 4 | 867 | ||
GenBank mRNA | AK000405.1 | - | 4 | 802 | ||
AK293227.1 | - | 3 | 1029 | |||
BC043346.2 | - | 4 | 794 | 6058941 | ||
BC053589.1 | - | 4 | 828 | 5246329 |