Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Champ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107329.1 | NP_001100799.1 | 2 | 2421 | ||
XM_006253396.1 | XP_006253458.1 | 2 | 2412 | |||
GenBank mRNA | BC168662.1 | - | 2 | 2418 | 7930141 |