Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RIPK1 SIRNA Assays ›
Gene Symbol:
Ripk1
Gene Name:
receptor (TNFRSF)-interacting serine-threonine kinase 1
Gene Aliases:
-
Chromosome Location:
Chr.17: 31537591 - 31569904 on Build Rnor_6.0
UniGene ID:
Rn.7572
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
s157818
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Ripk1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001107350.1
NP_001100820.1
4
744
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of protein phosphorylation
apoptotic process
positive regulation of cell death
positive regulation of interleukin-8 production
positive regulation of tumor necrosis factor production
response to tumor necrosis factor
peptidyl-serine autophosphorylation
positive regulation of phosphorylation
positive regulation of apoptotic process
positive regulation of programmed cell death
positive regulation of I-kappaB kinase/NF-kappaB signaling
negative regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of MAPK cascade
cellular protein catabolic process
positive regulation of macrophage differentiation
positive regulation of transcription from RNA polymerase II promoter
positive regulation of JNK cascade
protein autophosphorylation
positive regulation of NF-kappaB transcription factor activity
protein homooligomerization
protein heterooligomerization
positive regulation of transferase activity
positive regulation of necroptotic process
T cell apoptotic process
necroptotic process
regulation of ATP:ADP antiporter activity
cellular response to tumor necrosis factor
cellular response to growth factor stimulus
extrinsic apoptotic signaling pathway
ripoptosome assembly
necroptotic signaling pathway
ripoptosome assembly involved in necroptotic process
amyloid fibril formation
regulation of reactive oxygen species metabolic process
negative regulation of extrinsic apoptotic signaling pathway
positive regulation of extrinsic apoptotic signaling pathway
negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
protein kinase activity
protein serine/threonine kinase activity
death receptor binding
protein binding
ATP binding
ubiquitin protein ligase binding
protein complex binding
identical protein binding
death domain binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0