Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COLQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005677.3 | NP_005668.2 | 17 | 2102 | ||
NM_080538.2 | NP_536799.1 | 17 | 2231 | |||
NM_080539.3 | NP_536800.2 | 16 | 2000 | |||
GenBank mRNA | AF057036.1 | - | 17 | 2102 | ||
AK128401.1 | - | 17 | 2231 |