Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akr1c19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100576.1 | NP_001094046.1 | 4 | 507 | ||
XM_008771866.1 | XP_008770088.1 | 3 | 1673 | |||
XM_008771867.1 | XP_008770089.1 | 4 | 1368 |