Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006254819.2 | XP_006254881.1 | 12 | 1714 | ||
XM_006254820.2 | XP_006254882.1 | 11 | 1580 | |||
XM_006254821.2 | XP_006254883.1 | 12 | 1673 | |||
XM_006254822.2 | XP_006254884.1 | 11 | 1539 | |||
XM_006254823.2 | XP_006254885.1 | 12 | 1713 | |||
XM_006254824.2 | XP_006254886.1 | 12 | 1713 | |||
XM_006254825.2 | XP_006254887.1 | 12 | 1712 |