Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100974.1 | NP_001094444.1 | 3, 4 | 430 | ||
XM_006254807.2 | XP_006254869.1 | 3, 4 | 489 | |||
XM_006254808.2 | XP_006254870.1 | 3, 4 | 2429 | |||
XM_006254809.2 | XP_006254871.1 | 3, 4 | 396 | |||
XM_006254811.2 | XP_006254873.1 | 3, 4 | 525 | |||
GenBank mRNA | AB199794.1 | - | 3, 4 | 430 |