Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcerg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107390.2 | NP_001100860.2 | 4 | 539 | ||
XM_006254673.2 | XP_006254735.1 | 4 | 542 | |||
XM_006254674.2 | XP_006254736.1 | 4 | 542 | |||
XM_006254675.2 | XP_006254737.1 | 4 | 542 | |||
GenBank mRNA | BC168862.1 | - | 4 | 511 | 7444403 |