Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csnk2a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107409.1 | NP_001100879.1 | 9 | 788 | ||
XM_006255082.1 | XP_006255144.1 | 9 | 788 | |||
GenBank mRNA | BC168200.1 | - | 9 | 804 | 7114331 |