Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfat5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107425.1 | NP_001100895.1 | 6 | 1861 | ||
XM_003751872.3 | XP_003751920.1 | 5 | 1522 | |||
XM_006255553.2 | XP_006255615.1 | 4 | 4284 | |||
XM_006255554.2 | XP_006255616.1 | 4 | 3865 | |||
XM_006255555.2 | XP_006255617.1 | 5 | 532 | |||
XM_006255556.2 | XP_006255618.1 | 4 | 469 | |||
XM_008772503.1 | XP_008770725.1 | 6 | 1576 | |||
XM_008772504.1 | XP_008770726.1 | 4 | 1403 | |||
XM_008772505.1 | XP_008770727.1 | 3 | 4317 |