Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chst5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107431.1 | NP_001100901.1 | 2 | 923 | ||
XM_006255632.2 | XP_006255694.1 | 3 | 1067 | |||
XM_006255633.2 | XP_006255695.1 | 3 | 1701 | |||
GenBank mRNA | FQ212819.1 | - | 3 | 1067 | ||
FQ213398.1 | - | 2 | 957 |