Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004308.3 | NP_004299.1 | 6 | 663 | ||
XM_011520095.1 | XP_011518397.1 | 6 | 1100 | |||
XM_017017720.1 | XP_016873209.1 | 4 | 416 | |||
GenBank mRNA | AK298562.1 | - | 6 | 610 | ||
BC018118.1 | - | 6 | 568 | 3896574 | ||
U02570.1 | - | 6 | 570 | |||
U62794.1 | - | 1 | 187 | |||
Z23024.1 | - | 6 | 597 |