Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr126 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006227647.2 | XP_006227709.1 | 21 | 3492 | ||
XM_218313.8 | XP_218313.7 | 22 | 3577 |
Set Membership: |
G-protein coupled receptor |