Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spred3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173344.1 | NP_001166815.1 | 5 | 1715 | ||
XM_008759144.1 | XP_008757366.1 | 6 | 1353 | |||
XM_008759145.1 | XP_008757367.1 | 6 | 1300 | |||
XM_008759146.1 | XP_008757368.1 | 6 | 1422 | |||
GenBank mRNA | BC166515.1 | - | 5 | 1715 | 7131550 |