Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrfn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107502.1 | NP_001100972.1 | 1 | 1388 | ||
XM_006228771.2 | XP_006228833.1 | 2 | 2403 | |||
GenBank mRNA | BC158862.1 | - | 2 | 1703 | 7933283 |