Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klk8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107509.1 | NP_001100979.1 | 5 | 1216 | ||
XM_006229049.2 | XP_006229111.1 | 6 | 817 | |||
GenBank mRNA | AJ005641.1 | - | 5 | 884 |