Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tead2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107512.1 | NP_001100982.1 | 4 | 453 | ||
XM_006229069.1 | XP_006229131.1 | 4 | 408 | |||
XM_006229070.2 | XP_006229132.1 | 4 | 410 | |||
XM_008759378.1 | XP_008757600.1 | 4 | 492 | |||
GenBank mRNA | BC168947.1 | - | 4 | 402 | 9027778 |