Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcnxl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006230947.2 | XP_006231009.1 | 27 | 5044 | ||
XM_006230948.2 | XP_006231010.1 | 27 | 5041 | |||
XM_006230950.2 | XP_006231012.1 | 27 | 5014 | |||
XM_006230952.2 | XP_006231014.1 | 27 | 4975 | |||
XM_219515.8 | XP_219515.4 | 27 | 5005 |