Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRPX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170750.1 | NP_001164221.1 | 6 | 911 | ||
NM_001170751.1 | NP_001164222.1 | 6 | 794 | |||
NM_001170752.1 | NP_001164223.1 | 7 | 971 | |||
NM_006307.4 | NP_006298.1 | 7 | 971 | |||
XM_017029893.1 | XP_016885382.1 | 7 | 910 | |||
GenBank mRNA | AK125542.1 | - | 6 | 892 | ||
AK289430.1 | - | 7 | 918 | |||
AK293127.1 | - | 7 | 948 | |||
AK298802.1 | - | 6 | 730 | |||
BC020684.1 | - | 7 | 926 | 4731946 | ||
EU668359.1 | - | 7 | 871 | |||
U61374.1 | - | 7 | 871 | |||
U78093.1 | - | 7 | 878 |