Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRPX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170750.1 | NP_001164221.1 | 9 | 1324 | ||
NM_001170751.1 | NP_001164222.1 | 9 | 1207 | |||
NM_001170752.1 | NP_001164223.1 | 9 | 1262 | |||
NM_006307.4 | NP_006298.1 | 10 | 1384 | |||
GenBank mRNA | AK125542.1 | - | 9 | 1305 | ||
AK289430.1 | - | 10 | 1331 | |||
AK293127.1 | - | 9 | 1239 | |||
AK298802.1 | - | 9 | 1143 | |||
BC020684.1 | - | 10 | 1339 | 4731946 | ||
EU668359.1 | - | 10 | 1284 | |||
U61374.1 | - | 10 | 1284 | |||
U78093.1 | - | 10 | 1291 |