Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Otud7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008759566.1 | XP_008757788.1 | 3 | 621 | ||
XM_008759567.1 | XP_008757789.1 | 3 | 902 | |||
XM_008759568.1 | XP_008757790.1 | 3 | 621 |