Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kank1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037197.2 | NP_001032274.2 | 11 | 3996 | ||
XM_006231212.2 | XP_006231274.1 | 9 | 4939 | |||
XM_006231213.2 | XP_006231275.1 | 9 | 6620 | |||
XM_006231214.2 | XP_006231276.1 | 11 | 4113 | |||
XM_006231215.2 | XP_006231277.1 | 10 | 4024 | |||
XM_006231216.2 | XP_006231278.1 | 11 | 6547 | |||
XM_006231218.2 | XP_006231280.1 | 8 | 4687 | |||
XM_006231219.1 | XP_006231281.1 | 9 | 3937 | |||
XM_006231220.1 | XP_006231282.1 | 9 | 3860 | |||
XM_008760305.1 | XP_008758527.1 | 11 | 4130 | |||
GenBank mRNA | BC107446.1 | - | 5 | 645 |