Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpeb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760379.1 | XP_008758601.1 | 4 | 2560 | ||
XM_008760380.1 | XP_008758602.1 | 4 | 2491 | |||
XM_008760381.1 | XP_008758603.1 | 4 | 2052 | |||
XM_008760382.1 | XP_008758604.1 | 3 | 1271 | |||
XM_008760383.1 | XP_008758605.1 | 4 | 1406 | |||
XM_008760384.1 | XP_008758606.1 | 4 | 2560 |