Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: L3mbtl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003748666.3 | XP_003748714.1 | 4 | 616 | ||
XM_006227693.2 | XP_006227755.1 | 3 | 2272 | |||
XM_006227695.2 | XP_006227757.1 | 4 | 330 | |||
XM_006227696.2 | XP_006227758.1 | 3 | 249 | |||
XM_006227697.2 | XP_006227759.1 | 3 | 535 | |||
XM_006227698.2 | XP_006227760.1 | 3 | 2272 | |||
XM_006227699.2 | XP_006227761.1 | 2 | 95 | |||
XM_008758644.1 | XP_008756866.1 | 3 | 1451 |