Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epb41l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006227709.1 | XP_006227771.1 | 9 | 1526 | ||
XM_008758646.1 | XP_008756868.1 | 9 | 1708 | |||
XM_008758647.1 | XP_008756869.1 | 9 | 1824 | |||
XM_008758648.1 | XP_008756870.1 | 9 | 1429 | |||
XM_008758649.1 | XP_008756871.1 | 10 | 1664 | |||
XM_008758650.1 | XP_008756872.1 | 9 | 4511 | |||
GenBank mRNA | FQ212032.1 | - | 10 | 1622 |