Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RT1-M4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168343.1 | NP_001161815.1 | 3 | 368 | ||
XM_006255871.2 | XP_006255933.1 | 3 | 566 |