Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ascc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277057.1 | NP_001263986.1 | 4 | 921 | ||
XM_006256602.2 | XP_006256664.1 | 5 | 1019 | |||
XM_006256603.2 | XP_006256665.1 | 6 | 1127 | |||
XM_008772992.1 | XP_008771214.1 | 7 | 1196 | |||
XM_008772993.1 | XP_008771215.1 | 5 | 1040 |