Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sept10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014033.1 | NP_001014055.1 | 8 | 1086 | ||
XM_006256415.2 | XP_006256477.1 | 9 | 1311 | |||
XM_006256417.2 | XP_006256479.1 | 8 | 1304 | |||
XM_006256418.2 | XP_006256480.1 | 9 | 1310 | |||
XM_008772886.1 | XP_008771108.1 | 8 | 1589 | |||
GenBank mRNA | BC087157.1 | - | 8 | 1086 | 7133379 |