Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077647.1 | NP_001071115.1 | 21 | 2223 | ||
GenBank mRNA | GQ243745.1 | - | 20 | 2003 | ||
JQ013737.1 | - | 20 | 2011 | |||
S75275.1 | - | 21 | 2223 |