Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHX16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164239.1 | NP_001157711.1 | 2 | 178 | ||
NM_003587.4 | NP_003578.2 | 2 | 499 | |||
XM_011514940.2 | XP_011513242.1 | 1 | 31 | |||
GenBank mRNA | AB001601.1 | - | 2 | 426 | ||
AK302721.1 | - | 2 | 172 | |||
AK312264.1 | - | 2 | 438 | |||
BC002789.2 | - | 2 | 422 | |||
BC008825.2 | - | 2 | 450 | 4100320 | ||
BC009392.2 | - | 2 | 450 | 4136069 | ||
DB072495.1 | - | 2 | 500 |