Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubqln4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107688.1 | NP_001101158.1 | 6 | 1021 | ||
XM_006232642.1 | XP_006232704.1 | 5 | 815 |