Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CUL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257198.1 | NP_001244127.1 | 2 | 211 | ||
NM_003590.4 | NP_003581.1 | 2 | 528 | |||
XM_006712800.2 | XP_006712863.2 | 2 | 201 | |||
XM_011511994.2 | XP_011510296.1 | 2 | 199 | |||
XM_011511995.1 | XP_011510297.1 | 2 | 113 | |||
XM_011511996.2 | XP_011510298.1 | 1 | 51 | |||
GenBank mRNA | AB014517.2 | - | 2 | 456 | ||
AF062537.1 | - | 2 | 229 | |||
AF064087.1 | - | 2 | 221 | |||
AK291151.1 | - | 2 | 178 | |||
AY337761.1 | - | 2 | 674 | |||
BC039598.1 | - | 2 | 473 | 5784147 | ||
BC092409.1 | - | 2 | 473 | 6168545 | ||
BQ688156.1 | - | 528 |