Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pbxip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100976.1 | NP_001094446.1 | 4, 5 | 444 | ||
XM_006232668.1 | XP_006232730.1 | 5, 6 | 571 | |||
XM_006232669.1 | XP_006232731.1 | 5, 6 | 550 | |||
XM_008761174.1 | XP_008759396.1 | 5, 6 | 595 | |||
XM_008761175.1 | XP_008759397.1 | 5, 6 | 595 | |||
GenBank mRNA | BC085776.1 | - | 5, 6 | 459 | ||
BC129100.1 | - | 4, 5 | 444 | 7098205 |