Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pbxip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100976.1 | NP_001094446.1 | 9 | 1016 | ||
XM_006232668.1 | XP_006232730.1 | 10 | 1143 | |||
XM_006232669.1 | XP_006232731.1 | 10 | 1122 | |||
XM_008761174.1 | XP_008759396.1 | 10 | 1167 | |||
XM_008761175.1 | XP_008759397.1 | 9 | 1038 | |||
GenBank mRNA | BC085776.1 | - | 8 | 1905 | ||
BC129100.1 | - | 9 | 1016 | 7098205 |