Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekho1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025119.1 | NP_001020290.1 | 2 | 242 | ||
GenBank mRNA | BC091420.1 | - | 2 | 242 | 7365370 | |
FQ230444.1 | - | 1 | 51 | |||
FQ230757.1 | - | 1 | 92 |