Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polr3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012081.1 | NP_001012081.1 | 3 | 640 | ||
XM_006232967.1 | XP_006233029.1 | 3 | 847 | |||
XM_008761310.1 | XP_008759532.1 | 3 | 570 | |||
XM_008761311.1 | XP_008759533.1 | 3 | 847 | |||
XM_008761312.1 | XP_008759534.1 | 3 | 847 | |||
GenBank mRNA | BC083667.1 | - | 3 | 640 | 7135563 |