Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ngf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277055.1 | NP_001263984.1 | 3 | 191 | ||
XM_006233052.2 | XP_006233114.1 | 3 | 287 | |||
XM_006233053.2 | XP_006233115.1 | 3 | 222 | |||
GenBank mRNA | FQ232560.1 | - | 3 | 191 |