Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPST1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003596.3 | NP_003587.1 | 2 | 848 | ||
XM_005250642.2 | XP_005250699.1 | 2 | 844 | |||
XM_011516634.2 | XP_011514936.1 | 2 | 844 | |||
XM_011516635.2 | XP_011514937.1 | 2 | 844 | |||
XM_017012724.1 | XP_016868213.1 | 3 | 931 | |||
XM_017012725.1 | XP_016868214.1 | 3 | 931 | |||
XM_017012726.1 | XP_016868215.1 | 2 | 844 | |||
XM_017012727.1 | XP_016868216.1 | 2 | 844 | |||
GenBank mRNA | AF038009.1 | - | 1 | 577 | ||
AK313098.1 | - | 2 | 786 | |||
BC013188.1 | - | 2 | 823 | 3918922 | ||
CR542060.1 | - | 1 | 496 | |||
CR542080.1 | - | 1 | 496 |