Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEAD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256658.1 | NP_001243587.1 | 8, 9 | 876 | ||
NM_001256659.1 | NP_001243588.1 | 8, 9 | 832 | |||
NM_001256660.1 | NP_001243589.1 | 9, 10 | 875 | |||
NM_001256661.1 | NP_001243590.1 | 9, 10 | 850 | |||
NM_001256662.1 | NP_001243591.1 | 8, 9 | 606 | |||
NM_003598.1 | NP_003589.1 | 8, 9 | 823 | |||
XM_005259334.3 | XP_005259391.1 | 6, 7 | 987 | |||
XM_006723424.1 | XP_006723487.1 | 8, 9 | 1107 | |||
XM_006723428.2 | XP_006723491.1 | 7, 8 | 1095 | |||
XM_006723429.2 | XP_006723492.1 | 3, 4 | 308 | |||
XM_011527398.1 | XP_011525700.1 | 8, 9 | 1116 | |||
XM_011527399.2 | XP_011525701.1 | 9, 10 | 839 | |||
XM_011527400.2 | XP_011525702.1 | 9, 10 | 809 | |||
XM_011527401.1 | XP_011525703.1 | 9, 10 | 782 | |||
XM_011527402.2 | XP_011525704.1 | 9, 10 | 784 | |||
XM_011527403.1 | XP_011525705.1 | 8, 9 | 808 | |||
XM_011527404.1 | XP_011525706.1 | 8, 9 | 1116 | |||
GenBank mRNA | AK093236.1 | - | 7, 8 | 1104 | ||
AK290736.1 | - | 8, 9 | 829 | |||
AK300241.1 | - | 8, 9 | 590 | |||
AL832240.1 | - | 9, 10 | 800 | |||
BC007556.1 | - | 8, 9 | 823 | 2967735 | ||
BC018803.1 | - | 9, 10 | 850 | |||
BC051301.1 | - | 8, 9 | 826 | 4843913 | ||
X94440.1 | - | 7, 8 | 639 |