Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IRS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003604.2 | NP_003595.1 | 1 | 363 | ||
XM_005262220.2 | XP_005262277.1 | 1 | 570 | |||
XM_006724713.3 | XP_006724776.1 | 1 | 570 | |||
XM_011531061.1 | XP_011529363.1 | 1 | 570 | |||
GenBank mRNA | AF007567.1 | - | 1 | 408 |