Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC691920 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_001080099.5 | XP_001080099.4 | 5799 | |||
XM_006234815.2 | XP_006234877.1 | 5799 | ||||
XM_006234817.2 | XP_006234879.1 | 4531 |