Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif16b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107783.2 | NP_001101253.2 | 23 | 3640 | ||
XM_006235112.2 | XP_006235174.1 | 24 | 3808 | |||
XM_006235113.2 | XP_006235175.1 | 24 | 3808 | |||
XM_008762267.1 | XP_008760489.1 | 24 | 3808 | |||
XM_008762268.1 | XP_008760490.1 | 24 | 3808 |