Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hm13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107789.1 | NP_001101259.1 | 5 | 584 | ||
XM_006235268.1 | XP_006235330.1 | 5 | 595 | |||
XM_006235269.1 | XP_006235331.1 | 5 | 595 | |||
XM_008762330.1 | XP_008760552.1 | 5 | 595 |