Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPFIBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003621.3 | NP_003612.2 | 2, 3 | 454 | ||
XM_005253177.2 | XP_005253234.1 | 2, 3 | 449 | |||
XM_006718349.2 | XP_006718412.1 | 2, 3 | 449 | |||
XM_006718350.2 | XP_006718413.1 | 2, 3 | 449 | |||
XM_011520413.2 | XP_011518715.1 | 3, 4 | 437 | |||
XM_011520414.2 | XP_011518716.1 | 2, 3 | 164 | |||
XM_011520415.2 | XP_011518717.1 | 2, 3 | 449 | |||
XM_011520416.2 | XP_011518718.1 | 2, 3 | 449 | |||
XM_017018443.1 | XP_016873932.1 | 3, 4 | 437 | |||
GenBank mRNA | AK308398.1 | - | 2, 3 | 220 | ||
AL834426.1 | - | 1, 2 | 83 | |||
BC021714.1 | - | 2, 3 | 402 | 4826610 | ||
BG720018.1 | - | 3, 4 | 611 | |||
DA339729.1 | - | 2, 3 | 449 |