Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPFIA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304331.1 | NP_001291260.1 | 18 | 2664 | ||
NM_001304332.1 | NP_001291261.1 | 18 | 2664 | |||
XM_006711586.2 | XP_006711649.1 | 18 | 2694 | |||
XM_011510070.2 | XP_011508372.1 | 6 | 723 | |||
XM_017002606.1 | XP_016858095.1 | 18 | 2694 | |||
XM_017002607.1 | XP_016858096.1 | 17 | 2559 | |||
XM_017002608.1 | XP_016858097.1 | 18 | 2694 | |||
XM_017002609.1 | XP_016858098.1 | 17 | 2559 | |||
GenBank mRNA | AF034801.1 | - | 3 | 215 | ||
AK126624.1 | - | 5 | 1232 | |||
AK295757.1 | - | 12 | 1392 | |||
BC132923.1 | - | 5 | 1174 | 40146945 | ||
BC132925.1 | - | 5 | 1174 | 40146947 | ||
BC144262.1 | - | 5 | 1174 | 9052782 | ||
EF428334.1 | - | 23 | 2531 |