Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cables2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006235831.2 | XP_006235893.2 | 8 | 1067 | ||
XM_008762585.1 | XP_008760807.1 | 8 | 1076 | |||
XM_008762586.1 | XP_008760808.1 | 8 | 1067 | |||
XM_008762588.1 | XP_008760810.1 | 7 | 1004 | |||
XM_008762589.1 | XP_008760811.1 | 8 | 1076 | |||
XM_008762590.1 | XP_008760812.1 | 8 | 1076 | |||
XM_008762591.1 | XP_008760813.1 | 8 | 1076 |