Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107824.2 | NP_001101294.2 | 10 | 959 | ||
XM_006233799.2 | XP_006233861.1 | 9 | 1131 | |||
GenBank mRNA | BC160917.1 | - | 10 | 959 | 7457829 |