Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ophn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107848.1 | NP_001101318.1 | 12 | 1441 | ||
XM_006257072.2 | XP_006257134.1 | 11 | 1882 |