Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcmtd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257345.1 | NP_001244274.1 | 1 | 195 | ||
XM_008763511.1 | XP_008761733.1 | 3 | 1350 | |||
XM_008763512.1 | XP_008761734.1 | 2 | 1240 | |||
XM_008763513.1 | XP_008761735.1 | 3 | 1350 | |||
XM_008763514.1 | XP_008761736.1 | 2 | 1240 |